Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185095.1 | NP_001172024.1 | 7 | 669 | ||
NM_001185096.1 | NP_001172025.1 | 6 | 520 | |||
NM_031426.3 | NP_113614.1 | 6 | 591 | |||
NR_033701.1 | 6 | 613 | ||||
XM_017015177.1 | XP_016870666.1 | 5 | 525 | |||
GenBank mRNA | AK022845.1 | - | 7 | 536 | ||
AK057789.1 | - | 7 | 559 | |||
AK091912.1 | - | 6 | 504 | |||
AK128526.1 | - | 7 | 614 | |||
AK300473.1 | - | 5 | 530 | |||
AK314600.1 | - | 6 | 538 | |||
AL136566.1 | - | 6 | 525 | |||
AL833896.1 | - | 6 | 525 | |||
AY359067.1 | - | 6 | 542 | |||
BC021253.2 | - | 6 | 445 | 3504509 | ||
BI766795.1 | - | 6 | 564 |