Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305017.1 | NP_001291946.1 | 3 | 230 | ||
NM_130897.2 | NP_570967.1 | 3 | 237 | |||
NR_130942.1 | 3 | 778 | ||||
NR_130943.1 | 4 | 327 | ||||
XM_006721295.3 | XP_006721358.1 | 3 | 434 | |||
XM_006721296.3 | XP_006721359.1 | 3 | 187 | |||
XM_011523370.2 | XP_011521672.1 | 3 | 194 | |||
GenBank mRNA | AF125108.1 | - | 3 | 232 | ||
BC054892.1 | - | 4 | 258 | 6619254 | ||
CD676360.1 | - | 3 | 230 | |||
DB458006.1 | - | 3 | 199 | |||
HY018370.1 | - | 3 | 238 | |||
HY123054.1 | - | 3 | 220 | |||
HY201608.1 | - | 2 | 427 |