Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033678.3 | NP_001028850.2 | 8 | 961 | ||
NM_001160389.1 | NP_001153861.1 | 8 | 967 | |||
NM_001160390.1 | NP_001153862.1 | 8 | 949 | |||
NM_001160392.1 | NP_001153864.1 | 7 | 850 | |||
NM_001160393.1 | NP_001153865.1 | 7 | 759 | |||
NM_031472.3 | NP_113660.1 | 7 | 877 | |||
XM_005274345.4 | XP_005274402.1 | 8 | 884 | |||
XM_005274346.4 | XP_005274403.1 | 8 | 843 | |||
XM_005274347.4 | XP_005274404.1 | 7 | 760 | |||
XM_006718706.3 | XP_006718769.1 | 8 | 846 | |||
XM_006718707.3 | XP_006718770.1 | 8 | 793 | |||
XM_017018400.1 | XP_016873889.1 | 7 | 760 | |||
XM_017018401.1 | XP_016873890.1 | 6 | 721 | |||
GenBank mRNA | AK026364.1 | - | 6 | 1671 | ||
AY211494.1 | - | 8 | 917 | |||
BC005133.2 | - | 7 | 773 | 3836683 | ||
BM982538.1 | - | 1 | 57 | |||
BQ672032.1 | - | 6 | 664 |