Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTTY14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001543.3 | 1 | 246 | |||
NR_125733.1 | 1 | 246 | ||||
NR_125734.1 | 1 | 246 | ||||
NR_125735.1 | 1 | 246 | ||||
GenBank mRNA | BI824313.1 | - | 1 | 40 | ||
DA411985.1 | - | 1 | 246 | |||
DB155066.1 | - | 1 | 239 |