Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD3NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032016.3 | NP_114405.1 | 6 | 777 | ||
XM_005249880.1 | XP_005249937.1 | 7 | 791 | |||
XM_005249881.2 | XP_005249938.1 | 5 | 718 | |||
XM_011515572.1 | XP_011513874.1 | 6 | 712 | |||
XM_017012692.1 | XP_016868181.1 | 6 | 810 | |||
XM_017012693.1 | XP_016868182.1 | 7 | 741 | |||
XM_017012694.1 | XP_016868183.1 | 6 | 772 | |||
XM_017012695.1 | XP_016868184.1 | 6 | 737 | |||
XM_017012696.1 | XP_016868185.1 | 6 | 884 | |||
XM_017012697.1 | XP_016868186.1 | 5 | 620 | |||
XM_017012698.1 | XP_016868187.1 | 5 | 756 | |||
GenBank mRNA | AJ492267.1 | - | 6 | 652 | ||
AK091579.1 | - | 4 | 2691 | |||
AK315404.1 | - | 6 | 746 | |||
AY358645.1 | - | 6 | 603 | |||
BC003074.2 | - | 5 | 562 | 3505985 | ||
BC005959.1 | - | 6 | 597 | 4048871 |