Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032172.2 | NP_115548.1 | 14, 15 | 2320 | ||
XM_006715790.1 | XP_006715853.1 | 14, 15 | 2267 | |||
XM_006715791.2 | XP_006715854.1 | 17, 18 | 2773 | |||
XM_011515573.1 | XP_011513875.1 | 18, 19 | 2832 | |||
XM_011515574.1 | XP_011513876.1 | 15, 16 | 2356 | |||
XM_011515577.1 | XP_011513879.1 | 13, 14 | 1985 | |||
XM_011515578.1 | XP_011513880.1 | 15, 16 | 2407 | |||
GenBank mRNA | AJ601395.1 | - | 13, 14 | 2197 | ||
AK022759.1 | - | 14, 15 | 2264 | |||
AK301527.1 | - | 12, 13 | 1792 | |||
AY618868.1 | - | 14, 15 | 2264 | |||
BC060846.1 | - | 14, 15 | 2259 | |||
BC132862.1 | - | 13, 14 | 2217 | 40146884 | ||
BC136656.1 | - | 12, 13 | 3150 |