Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032193.3 | NP_115569.2 | 2, 3 | 526 | ||
GenBank mRNA | AF312034.1 | - | 2, 3 | 365 | ||
AF346606.1 | - | 2, 3 | 526 | |||
AK024627.1 | - | 2, 3 | 383 | |||
AK225131.1 | - | 2, 3 | 383 | |||
AK297660.1 | - | 2, 3 | 296 | |||
BC023588.2 | - | 2, 3 | 406 | 4763204 | ||
BM921525.1 | - | 2, 3 | 395 | |||
L08647.1 | - | 1, 2 | 255 |