Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290137.1 | NP_001277066.1 | 9 | 1242 | ||
NM_001290139.1 | NP_001277068.1 | 9 | 1312 | |||
NM_001323635.1 | NP_001310564.1 | 9 | 1347 | |||
NM_001323636.1 | NP_001310565.1 | 8 | 1267 | |||
NM_032304.3 | NP_115680.1 | 8 | 1273 | |||
XM_005255632.1 | XP_005255689.1 | 8 | 1480 | |||
XM_011522712.1 | XP_011521014.1 | 8 | 1504 | |||
XM_017023774.1 | XP_016879263.1 | 8 | 1336 | |||
GenBank mRNA | AA489499.1 | - | 1 | 81 | ||
AK054841.1 | - | 6 | 1568 | |||
AK098340.1 | - | 3 | 2100 | |||
AW136334.1 | - | 1 | 113 | |||
BC004353.1 | - | 8 | 1248 | 3351990 | ||
BC015008.1 | - | 8 | 1201 | 4862387 | ||
BC033796.1 | - | 7 | 1602 | 5173915 | ||
BC036747.1 | - | 7 | 1602 | |||
BI834617.1 | - | 4 | 582 | |||
BQ067068.1 | - | 4 | 560 | |||
BX090134.1 | - | 1 | 110 |