Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032315.2 | NP_115691.1 | 4 | 606 | ||
XM_005263503.3 | XP_005263560.1 | 4 | 275 | |||
XM_011542296.1 | XP_011540598.1 | 4 | 393 | |||
GenBank mRNA | AF495714.1 | - | 4 | 574 | ||
AJ880283.1 | - | 4 | 592 | |||
BC004991.1 | - | 4 | 576 | 2905957 | ||
BC073135.1 | - | 4 | 592 | 6172562 |