Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032339.4 | NP_115715.3 | 2, 3 | 262 | ||
XM_005257736.3 | XP_005257793.1 | 2, 3 | 241 | |||
GenBank mRNA | AF490253.1 | - | 2, 3 | 194 | ||
AJ308025.1 | - | 2, 3 | 283 | |||
AL554581.3 | - | 2, 3 | 484 | |||
AY508814.1 | - | 2, 3 | 262 | |||
BC006006.1 | - | 2, 3 | 201 | 4283597 | ||
BC106016.1 | - | 1, 2 | 330 | |||
BX646129.1 | - | 2, 3 | 235 |