Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032340.3 | NP_115716.1 | 4 | 424 | ||
GenBank mRNA | AF086234.1 | - | 4 | 346 | ||
AK127468.1 | - | 1 | 983 | |||
AK311835.1 | - | 4 | 373 | |||
AY568085.1 | - | 4 | 400 | |||
BC006007.1 | - | 4 | 400 | 4284544 | ||
BI604191.1 | - | 4 | 374 | |||
DA123098.1 | - | 4 | 424 |