Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032341.4 | NP_115717.3 | 3 | 499 | ||
GenBank mRNA | AK293006.1 | - | 3 | 499 | ||
BC006011.1 | - | 3 | 414 | 4296688 |