Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032412.3 | NP_115788.1 | 2 | 250 | ||
GenBank mRNA | AJ245877.1 | - | 2 | 140 | ||
AK225992.1 | - | 2 | 130 | |||
AK312045.1 | - | 2 | 126 | |||
BC013643.1 | - | 2 | 160 | 4152168 | ||
BC023982.1 | - | 2 | 167 | 4046171 | ||
BM919999.1 | - | 2 | 250 | |||
CA310907.1 | - | 2 | 672 |