Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032413.3 | NP_115789.1 | 1, 2 | 307 | ||
NM_197955.2 | NP_922946.1 | 2, 3 | 193 | |||
GenBank mRNA | AB026707.1 | - | 2, 3 | 87 | ||
AF228422.1 | - | 1, 2 | 228 | |||
AK026298.1 | - | 1, 2 | 225 | |||
BC021173.2 | - | 2, 3 | 113 | 5269504 | ||
BM312626.1 | - | 2, 3 | 157 | |||
CA865860.1 | - | 2, 3 | 199 | |||
DB447060.1 | - | 2, 3 | 195 |