Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSANTD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318747.1 | NP_001305676.1 | 2, 3 | 1056 | ||
NM_001318748.1 | NP_001305677.1 | 2, 3 | 1052 | |||
NM_001318749.1 | NP_001305678.1 | 2, 3 | 770 | |||
NM_001318750.1 | NP_001305679.1 | 2, 3 | 697 | |||
NM_032424.2 | NP_115800.1 | 2, 3 | 1930 | |||
XM_011543022.2 | XP_011541324.1 | 3, 4 | 1958 | |||
XM_017018416.1 | XP_016873905.1 | 3, 4 | 1068 | |||
XM_017018417.1 | XP_016873906.1 | 2, 3 | 655 | |||
GenBank mRNA | AB058729.1 | - | 2, 3 | 1870 | ||
AF253976.1 | - | 1, 2 | 120 | |||
AK027790.1 | - | 1, 2 | 90 | |||
AK293050.1 | - | 2, 3 | 726 | |||
AL834523.1 | - | 2, 3 | 620 | |||
BC041118.1 | - | 2, 3 | 627 | 5526792 |