Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032428.1 | NP_115804.1 | 11 | 1077 | ||
XM_006724709.2 | XP_006724772.1 | 5 | 552 | |||
XM_011531056.2 | XP_011529358.1 | 9 | 1053 | |||
XM_011531057.2 | XP_011529359.1 | 9 | 979 | |||
XM_011531058.2 | XP_011529360.1 | 8 | 891 | |||
XM_017029901.1 | XP_016885390.1 | 9 | 1298 | |||
XM_017029902.1 | XP_016885391.1 | 9 | 1298 |