Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYVN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032431.2 | NP_115807.1 | 14 | 1642 | ||
NM_172230.2 | NP_757385.1 | 14 | 1645 | |||
XM_011545303.2 | XP_011543605.1 | 14 | 2493 | |||
GenBank mRNA | AB058713.1 | - | 12 | 1437 | ||
AB085847.1 | - | 13 | 1548 | |||
AF317634.1 | - | 13 | 1548 | |||
AK074148.1 | - | 12 | 2055 | |||
AK126912.1 | - | 14 | 1646 | |||
AL834262.1 | - | 14 | 1642 | |||
BC030530.1 | - | 14 | 1586 | 5211742 |