Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYVN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032431.2 | NP_115807.1 | 10 | 1008 | ||
NM_172230.2 | NP_757385.1 | 10 | 1008 | |||
XM_011545303.2 | XP_011543605.1 | 10 | 1856 | |||
GenBank mRNA | AB058713.1 | - | 8 | 803 | ||
AB085847.1 | - | 9 | 914 | |||
AF317634.1 | - | 9 | 914 | |||
AK074148.1 | - | 9 | 1343 | |||
AK126912.1 | - | 10 | 1009 | |||
AL834262.1 | - | 10 | 1008 | |||
BC030530.1 | - | 10 | 949 | 5211742 |