Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011657.3 | NP_001011657.2 | 7 | 1987 | ||
NM_001282400.1 | NP_001269329.1 | 10 | 2543 | |||
NM_001282401.1 | NP_001269330.1 | 9 | 2490 | |||
XM_005262212.4 | XP_005262269.2 | 7 | 2004 | |||
XM_005262213.4 | XP_005262270.2 | 7 | 2001 | |||
XM_005262214.3 | XP_005262271.2 | 6 | 1935 | |||
XM_005262216.4 | XP_005262273.2 | 7 | 1706 | |||
XM_006724711.3 | XP_006724774.1 | 3 | 1466 | |||
XM_017029903.1 | XP_016885392.1 | 7 | 1703 | |||
XM_017029904.1 | XP_016885393.1 | 7 | 3756 | |||
GenBank mRNA | AB058692.1 | - | 2 | 4620 | ||
AK094141.1 | - | 10 | 2504 | |||
AK296520.1 | - | 9 | 2455 | |||
AL831883.1 | - | 1 | 354 | |||
BC140920.1 | - | 2 | 4586 | 9056732 | ||
BC152470.1 | - | 2 | 4620 | 8862607 |