Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145459.1 | NP_001138931.1 | 2 | 280 | ||
NM_001145460.1 | NP_001138932.1 | 3 | 634 | |||
NM_032495.5 | NP_115884.4 | 3 | 634 | |||
NM_139211.4 | NP_631957.1 | 2 | 236 | |||
NM_139212.3 | NP_631958.1 | 2 | 509 | |||
XM_006714052.2 | XP_006714115.1 | 3 | 629 | |||
XM_017008728.1 | XP_016864217.1 | 2 | 271 | |||
XM_017008729.1 | XP_016864218.1 | 2 | 153 | |||
XM_017008730.1 | XP_016864219.1 | 2 | 201 | |||
XM_017008731.1 | XP_016864220.1 | 2 | 236 | |||
XM_017008732.1 | XP_016864221.1 | 3 | 512 | |||
XM_017008733.1 | XP_016864222.1 | 2 | 76 | |||
XM_017008734.1 | XP_016864223.1 | 2 | 132 | |||
GenBank mRNA | AB059408.1 | - | 2 | 194 | ||
AB059409.1 | - | 2 | 77 | |||
AB059410.2 | - | 2 | 86 | |||
AF454763.1 | - | 2 | 99 | |||
AF492675.1 | - | 2 | 483 | |||
AF492676.1 | - | 3 | 215 | |||
AF492677.1 | - | 3 | 259 | |||
AF492678.1 | - | 3 | 259 | |||
AF492679.1 | - | 3 | 215 | |||
AF492680.1 | - | 2 | 235 | |||
AF492681.1 | - | 2 | 198 | |||
AK289707.1 | - | 2 | 178 | |||
AW955211.1 | - | 2 | 202 | |||
BC014225.2 | - | 1 | 349 | 4335211 | ||
CB996541.1 | - | 2 | 272 | |||
CK002567.1 | - | 2 | 112 | |||
DA512059.1 | - | 2 | 509 | |||
EB386856.1 | - | 2 | 230 |