Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027128.1 | 4 | 1263 | |||
GenBank mRNA | AY035381.1 | - | 2 | 555 | ||
BC101235.2 | - | 1 | 501 | |||
BC101236.2 | - | 1 | 501 | |||
BC101237.1 | - | 1 | 501 | |||
BC101238.2 | - | 1 | 501 | |||
BM674075.1 | - | 1 | 584 |