Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032517.4 | NP_115906.3 | 3, 4 | 486 | ||
XM_005252627.3 | XP_005252684.1 | 3, 4 | 496 | |||
GenBank mRNA | AI393331.1 | - | 2, 3 | 324 | ||
AY358694.1 | - | 3, 4 | 431 | |||
BC021730.2 | - | 3, 4 | 452 | 4825273 | ||
BP369317.1 | - | 3, 4 | 497 |