Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005366.1 | NP_001005366.1 | 7 | 1063 | ||
NM_032590.4 | NP_115979.3 | 7 | 853 | |||
XM_005253955.3 | XP_005254012.1 | 7 | 1063 | |||
XM_005253956.3 | XP_005254013.1 | 7 | 660 | |||
XM_011538867.2 | XP_011537169.1 | 7 | 2106 | |||
XM_011538868.2 | XP_011537170.1 | 7 | 2102 | |||
XM_011538869.2 | XP_011537171.1 | 7 | 2104 | |||
GenBank mRNA | AJ459424.1 | - | 7 | 825 | ||
AK127328.1 | - | 7 | 1063 | |||
AL133572.1 | - | 3 | 366 | |||
BC115379.1 | - | 6 | 713 | |||
BC115380.1 | - | 7 | 792 | 40029132 | ||
BX648928.1 | - | 7 | 855 |