Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128855.2 | NP_001122327.1 | 5, 6 | 770 | ||
NM_001195422.1 | NP_001182351.1 | 5, 6 | 763 | |||
NM_032620.3 | NP_116009.2 | 5, 6 | 770 | |||
NM_133644.3 | NP_598399.2 | 5 | 866 | |||
GenBank mRNA | AF360742.1 | - | 5, 6 | 732 | ||
AF360743.1 | - | 5, 6 | 732 | |||
AF360744.1 | - | 5, 6 | 712 | |||
AK027606.1 | - | 5, 6 | 717 | |||
AK091035.1 | - | 5 | 1571 | |||
AK291929.1 | - | 5, 6 | 714 | |||
AK297953.1 | - | 5, 6 | 745 | |||
AK298485.1 | - | 993 | ||||
AK311442.1 | - | 5, 6 | 833 | |||
AY078987.1 | - | 5 | 828 | |||
AY078988.1 | - | 5, 6 | 732 | |||
BC008924.2 | - | 3, 4 | 310 | |||
BC017207.2 | - | 5, 6 | 702 | 3634066 | ||
BC019261.1 | - | 5, 6 | 711 | 3501739 | ||
BM789389.1 | - | 2, 3 | 234 | |||
DA490131.1 | - | 1 | 374 |