Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032731.3 | NP_116120.1 | 1 | 211 | ||
GenBank mRNA | AF086210.1 | - | 1 | 62 | ||
AJ344101.1 | - | 1 | 106 | |||
AK291963.1 | - | 1 | 95 | |||
AK311853.1 | - | 1 | 65 | |||
BC006405.2 | - | 1 | 89 | 4298953 | ||
CA431010.1 | - | 4 | 454 |