Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282613.1 | NP_001269542.1 | 2 | 337 | ||
NM_001282614.1 | NP_001269543.1 | 4 | 599 | |||
NM_001282615.1 | NP_001269544.1 | 2 | 337 | |||
NM_001282616.1 | NP_001269545.1 | 3 | 432 | |||
NM_032735.2 | NP_116124.2 | 4 | 599 | |||
XM_006719256.3 | XP_006719319.1 | 3 | 387 | |||
XM_011537961.1 | XP_011536263.1 | 2 | 207 | |||
XM_017018865.1 | XP_016874354.1 | 3 | 387 | |||
GenBank mRNA | AF440758.1 | - | 4 | 544 | ||
AK096459.1 | - | 4 | 600 | |||
AY515706.1 | - | 3 | 372 | |||
BC006440.1 | - | 2 | 254 | |||
BC151138.1 | - | 2 | 206 | |||
BP225850.1 | - | 3 | 411 |