Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146314.1 | NP_001139786.1 | 2 | 802 | ||
NM_032750.2 | NP_116139.1 | 2 | 996 | |||
GenBank mRNA | AK075112.1 | - | 2 | 387 | ||
AK296599.1 | - | 2 | 182 | |||
AK298022.1 | - | 2 | 210 | |||
AK298813.1 | - | 2 | 193 | |||
BC007234.1 | - | 2 | 380 | 2823284 | ||
BC050650.1 | - | 2 | 194 | |||
BC056411.1 | - | 2 | 203 | 6048106 | ||
BC071931.1 | - | 3 | 615 | |||
DA315486.1 | - | 2 | 252 | |||
FJ224306.1 | - | 223 |