Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193308.1 | NP_001180237.1 | 14 | 1629 | ||
NM_032872.2 | NP_116261.1 | 14 | 1593 | |||
XM_005246022.3 | XP_005246079.1 | 14 | 1784 | |||
XM_006710990.1 | XP_006711053.1 | 14 | 1748 | |||
XM_006710991.1 | XP_006711054.1 | 13 | 1383 | |||
GenBank mRNA | AK027902.1 | - | 14 | 1563 | ||
AK074154.1 | - | 7 | 3676 | |||
AK096437.1 | - | 12 | 2190 | |||
AK097172.1 | - | 7 | 2346 | |||
AK126120.1 | - | 1 | 1903 | |||
AK293038.1 | - | 13 | 1895 | |||
AY037157.1 | - | 14 | 1556 | |||
BC009224.2 | - | 12 | 1160 | |||
BC015764.1 | - | 13 | 1372 | 4851468 | ||
BC035725.1 | - | 14 | 1536 | 5722683 |