Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032878.3 | NP_116267.3 | 1, 2 | 68 | ||
NM_198867.1 | NP_942567.1 | 1, 2 | 68 | |||
GenBank mRNA | AK304805.1 | - | 1, 2 | 42 | ||
AK310130.1 | - | 1, 2 | 42 | |||
AK310704.1 | - | 1, 2 | 65 | |||
AK314150.1 | - | 1, 2 | 47 | |||
AK315894.1 | - | 1, 2 | 36 | |||
BC007601.1 | - | 1, 2 | 37 | |||
BC105801.1 | - | 1, 2 | 40 |