Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYURF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032906.4 | NP_116295.1 | 2 | 411 | ||
GenBank mRNA | AK312082.1 | - | 2 | 386 | ||
BG706896.1 | - | 2 | 416 | |||
CV812938.1 | - | 2 | 347 |
Gene Symbol: PIGY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042616.2 | NP_001036081.1 | 2 | 411 | ||
GenBank mRNA | AB206972.1 | - | 2 | 383 | ||
BC007876.2 | - | 2 | 383 | 3947944 | ||
BI548853.1 | - | 2 | 416 |