Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDXK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056502.1 | - | 4 | 932 | ||
AK094630.1 | - | 1 | 538 | |||
BC008008.1 | - | 2 | 321 | 3528340 | ||
BC021550.1 | - | 2 | 321 | 4905448 | ||
BC041020.1 | - | 2 | 321 | 3528340 |
Gene Symbol: LOC105372824 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529811.1 | XP_011528113.1 | 1 | 211 |