Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033060.2 | NP_149049.1 | 1 | 199 | ||
GenBank mRNA | AJ406942.1 | - | 1 | 199 | ||
BC065372.1 | - | 2 | 696 | |||
BC103845.1 | - | 1 | 179 | 40002744 | ||
BC103846.2 | - | 1 | 179 | 40002745 |