Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271082.1 | NP_001258011.1 | 8 | 728 | ||
NM_033120.3 | NP_149111.1 | 8 | 728 | |||
XM_005248382.3 | XP_005248439.1 | 8 | 845 | |||
XM_005248383.3 | XP_005248440.1 | 8 | 845 | |||
XM_011514150.2 | XP_011512452.1 | 8 | 845 | |||
XM_017010000.1 | XP_016865489.1 | 3 | 1031 | |||
GenBank mRNA | AB062887.1 | - | 8 | 637 | ||
AF358137.1 | - | 8 | 649 | |||
BC004940.1 | - | 3 | 256 | |||
BC012176.1 | - | 8 | 728 | 4649935 |