Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025248.1 | XP_016880737.1 | 1 | 62 | ||
XM_017025250.1 | XP_016880739.1 | 1 | 62 |