Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033395.1 | NP_203753.1 | 15 | 4820 | ||
XM_005274366.1 | XP_005274423.1 | 15 | 4823 | |||
XM_005274367.2 | XP_005274424.1 | 15 | 4823 | |||
XM_005274368.2 | XP_005274425.1 | 15 | 4823 | |||
XM_011543047.1 | XP_011541349.1 | 15 | 4823 | |||
XM_011543048.1 | XP_011541350.1 | 15 | 4784 | |||
XM_011543049.1 | XP_011541351.1 | 15 | 4823 | |||
XM_011543050.2 | XP_011541352.1 | 11 | 5164 | |||
XM_011543053.2 | XP_011541355.1 | 9 | 3983 | |||
XM_011543054.2 | XP_011541356.1 | 4 | 3237 | |||
XM_017018470.1 | XP_016873959.1 | 15 | 4823 | |||
XM_017018471.1 | XP_016873960.1 | 15 | 4823 | |||
XM_017018472.1 | XP_016873961.1 | 15 | 4823 | |||
XM_017018473.1 | XP_016873962.1 | 15 | 4791 | |||
XM_017018474.1 | XP_016873963.1 | 11 | 4205 | |||
XM_017018475.1 | XP_016873964.1 | 15 | 4752 | |||
XM_017018476.1 | XP_016873965.1 | 9 | 3983 | |||
GenBank mRNA | AB051518.2 | - | 15 | 4759 |