Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANC1 SIRNA Assays ›
Gene Symbol:
TANC1
Gene Name:
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
Gene Aliases:
ROLSB, TANC
Chromosome Location:
Chr.2: 158968600 - 159232659 on Build GRCh38
UniGene ID:
Hs.61590
Species:
Human
siRNA ID
HSS150347
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145909.1
NP_001139381.1
10
1586
NM_033394.2
NP_203752.2
10
1610
XM_005246918.4
XP_005246975.1
9
1374
XM_006712810.3
XP_006712873.1
9
1497
XM_006712812.3
XP_006712875.1
9
1498
XM_006712814.3
XP_006712877.1
6
793
XM_011512048.2
XP_011510350.1
12
1788
XM_011512051.2
XP_011510353.1
11
1653
XM_011512053.2
XP_011510355.1
10
1543
XM_011512055.2
XP_011510357.1
10
1579
XM_011512058.2
XP_011510360.1
8
1285
XM_011512059.2
XP_011510361.1
6
1146
XM_011512062.2
XP_011510364.1
4
823
XM_017005141.1
XP_016860630.1
9
1402
XM_017005142.1
XP_016860631.1
11
2007
XM_017005143.1
XP_016860632.1
9
1692
XM_017005144.1
XP_016860633.1
9
1381
XM_017005145.1
XP_016860634.1
12
1822
XM_017005146.1
XP_016860635.1
11
1633
XM_017005147.1
XP_016860636.1
10
1582
XM_017005148.1
XP_016860637.1
11
1748
XM_017005149.1
XP_016860638.1
8
1300
XM_017005150.1
XP_016860639.1
8
1282
XM_017005151.1
XP_016860640.1
8
1141
XM_017005152.1
XP_016860641.1
8
1138
XM_017005153.1
XP_016860642.1
7
1087
XM_017005154.1
XP_016860643.1
10
1504
XM_017005155.1
XP_016860644.1
8
1234
XM_017005156.1
XP_016860645.1
6
823
XM_017005157.1
XP_016860646.1
7
1000
XM_017005158.1
XP_016860647.1
12
1751
GenBank mRNA
AB051515.1
-
4
687
BC037329.1
-
8
1291
BC150597.1
-
9
1427
9056967
BX648327.1
-
5
870
Back To Top
More Information
Gene Ontology Categories:
Function(s)
myoblast fusion
visual learning
dendritic spine maintenance
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-wqf8l:80/100.66.76.116:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline