Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033390.1 | NP_203748.1 | 6 | 2281 | ||
XM_005271715.3 | XP_005271772.1 | 6 | 2662 | |||
XM_011543055.2 | XP_011541357.1 | 6 | 2394 | |||
XM_017018477.1 | XP_016873966.1 | 6 | 6832 | |||
GenBank mRNA | AB051513.1 | - | 4 | 1454 | ||
AB096241.1 | - | 5 | 3018 | |||
AK295437.1 | - | 6 | 2282 | |||
AK299100.1 | - | 6 | 2314 | |||
AL832893.1 | - | 3 | 1134 |