Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002312.1 | 1 | 262 | |||
GenBank mRNA | CA413366.1 | - | 1 | 99 | ||
CV309337.1 | - | 1 | 180 | |||
X15624.1 | - | 1 | 261 |
Gene Symbol: PARP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020912.1 | XP_016876401.1 | 1 | 106 |