Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289034.1 | NP_001275963.1 | 2 | 565 | ||
NM_001289035.1 | NP_001275964.1 | 2 | 331 | |||
NM_138343.3 | NP_612352.1 | 2 | 331 | |||
NM_201521.2 | NP_958929.1 | 2 | 331 | |||
NM_201522.2 | NP_958930.1 | 3 | 747 | |||
NM_201523.2 | NP_958931.1 | 2 | 621 | |||
XM_011514989.1 | XP_011513291.1 | 2 | 680 | |||
XM_017011457.1 | XP_016866946.1 | 2 | 331 | |||
GenBank mRNA | AK056393.1 | - | 2 | 291 | ||
AK094102.1 | - | 2 | 587 | |||
AK127018.1 | - | 5 | 1565 | |||
AK127894.1 | - | 2 | 304 | |||
AK297433.1 | - | 2 | 294 | |||
BC012357.1 | - | 2 | 272 | |||
BC080637.1 | - | 2 | 271 | 6573021 | ||
BC103727.1 | - | 2 | 269 | 6157949 | ||
BQ688778.1 | - | 2 | 401 | |||
DA782235.1 | - | 2 | 331 |