Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289034.1 | NP_001275963.1 | 2 | 354 | ||
NM_001289035.1 | NP_001275964.1 | 2 | 120 | |||
NM_138343.3 | NP_612352.1 | 2 | 120 | |||
NM_201521.2 | NP_958929.1 | 2 | 120 | |||
NM_201522.2 | NP_958930.1 | 3 | 536 | |||
NM_201523.2 | NP_958931.1 | 2 | 410 | |||
XM_011514989.1 | XP_011513291.1 | 2 | 469 | |||
XM_017011457.1 | XP_016866946.1 | 2 | 120 | |||
GenBank mRNA | AK055293.1 | - | 3 | 498 | ||
AK056393.1 | - | 2 | 80 | |||
AK094102.1 | - | 2 | 376 | |||
AK127018.1 | - | 5 | 1354 | |||
AK127894.1 | - | 2 | 75 | |||
AK297433.1 | - | 2 | 83 | |||
BC012357.1 | - | 2 | 61 | |||
BC080637.1 | - | 2 | 60 | 6573021 | ||
BC103727.1 | - | 2 | 58 | 6157949 | ||
BQ688778.1 | - | 2 | 190 | |||
BX640717.1 | - | 2 | 317 | |||
DA104494.1 | - | 2 | 410 | |||
DA782235.1 | - | 2 | 120 | |||
DA835055.1 | - | 3 | 498 |