Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177401.4 | NP_796375.3 | 8 | 1869 | ||
XM_005259671.3 | XP_005259728.1 | 9 | 1998 | |||
XM_005259672.3 | XP_005259729.1 | 9 | 1996 | |||
GenBank mRNA | BC015089.2 | - | 2 | 473 | ||
BC094778.1 | - | 7 | 1763 | 30342357 |