Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052846.1 | NP_443078.1 | 4 | 1625 | ||
GenBank mRNA | AB089149.1 | - | 4 | 1625 | ||
AL137580.1 | - | 1 | 493 | |||
BC101043.1 | - | 4 | 1401 | 40007928 | ||
BC101044.2 | - | 4 | 1401 | |||
BC101045.1 | - | 4 | 1401 | 40007932 | ||
BC101046.1 | - | 4 | 835 |