Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEGF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271938.1 | NP_001258867.1 | 42 | 10958 | ||
NM_001410.2 | NP_001401.2 | 41 | 10757 | |||
GenBank mRNA | AF497566.1 | - | 1 | 168 | ||
AK026018.1 | - | 1 | 584 | |||
BC093649.1 | - | 1 | 194 | |||
BC111942.1 | - | 1 | 194 |