Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042632.2 | NP_001036097.1 | 4 | 625 | ||
NM_001042633.2 | NP_001036098.1 | 5 | 643 | |||
NM_033421.3 | NP_219489.1 | 4 | 632 | |||
NM_152897.2 | NP_690857.1 | 4 | 632 | |||
XM_005260608.4 | XP_005260665.1 | 2 | 707 | |||
XM_011529095.2 | XP_011527397.1 | 2 | 611 | |||
GenBank mRNA | AF258570.1 | - | 1 | 292 | ||
AF395845.1 | - | 4 | 543 | |||
AF523834.1 | - | 4 | 611 | |||
AK091359.1 | - | 3 | 647 | |||
AK095851.1 | - | 3 | 506 | |||
AK226120.1 | - | 3 | 156 | |||
AK293443.1 | - | 5 | 644 | |||
AK297622.1 | - | 4 | 566 | |||
AL591562.1 | - | 4 | 611 | |||
BC012904.1 | - | 6 | 863 | |||
BC019823.1 | - | 4 | 605 | 4453502 | ||
BM925584.1 | - | 5 | 629 | |||
KU178820.1 | - | 4 | 543 | |||
KU178821.1 | - | 5 | 554 |