Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133369.2 | NP_588610.2 | 8 | 1555 | ||
XM_006714927.1 | XP_006714990.1 | 8 | 1535 | |||
XM_006714928.1 | XP_006714991.1 | 9 | 1703 | |||
XM_011534686.1 | XP_011532988.1 | 9 | 1703 | |||
XM_011534687.2 | XP_011532989.1 | 9 | 1703 | |||
GenBank mRNA | AB075856.1 | - | 1 | 150 | ||
BC009333.2 | - | 2 | 461 | |||
BC122556.1 | - | 2 | 432 | |||
BC157824.1 | - | 8 | 1438 | 9057043 |