Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133369.2 | NP_588610.2 | 3 | 543 | ||
XM_006714927.1 | XP_006714990.1 | 3 | 523 | |||
XM_006714928.1 | XP_006714991.1 | 3 | 523 | |||
XM_011534686.1 | XP_011532988.1 | 3 | 523 | |||
XM_011534687.2 | XP_011532989.1 | 3 | 523 | |||
GenBank mRNA | AK131380.1 | - | 4 | 420 | ||
BC033727.1 | - | 4 | 671 | |||
BC157824.1 | - | 3 | 426 | 9057043 | ||
BI818609.1 | - | 3 | 354 |