Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138350.3 | NP_612359.2 | 5 | 569 | ||
XM_005263532.3 | XP_005263589.1 | 6 | 2079 | |||
XM_011542400.2 | XP_011540702.1 | 5 | 1653 | |||
XM_017002761.1 | XP_016858250.1 | 5 | 506 | |||
GenBank mRNA | AK096217.1 | - | 5 | 527 |