Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008801.1 | NP_001008801.1 | 4 | 2222 | ||
NM_001277120.1 | NP_001264049.1 | 5 | 2340 | |||
NR_102299.1 | 6 | 2595 | ||||
NR_102300.1 | 6 | 2576 | ||||
NR_102301.1 | 5 | 2477 | ||||
XM_017027443.1 | XP_016882932.1 | 5 | 2437 | |||
XM_017027444.1 | XP_016882933.1 | 6 | 2555 | |||
GenBank mRNA | AK023558.1 | - | 1 | 278 | ||
AK025501.1 | - | 1 | 860 | |||
AY442330.1 | - | 4 | 2219 | |||
AY442331.1 | - | 5 | 2337 | |||
BC063143.1 | - | 1 | 522 | |||
CR749381.1 | - | 1 | 1316 | |||
CR936599.1 | - | 4 | 2188 | |||
CR936695.1 | - | 4 | 2192 |