Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008801.1 | NP_001008801.1 | 4 | 2977 | ||
NM_001277120.1 | NP_001264049.1 | 5 | 3095 | |||
NR_102299.1 | 6 | 3350 | ||||
NR_102300.1 | 6 | 3331 | ||||
NR_102301.1 | 5 | 3232 | ||||
XM_017027443.1 | XP_016882932.1 | 5 | 3192 | |||
XM_017027444.1 | XP_016882933.1 | 6 | 3310 | |||
GenBank mRNA | AK023558.1 | - | 1 | 1033 | ||
AK025501.1 | - | 1 | 1615 | |||
AY442330.1 | - | 4 | 2974 | |||
AY442331.1 | - | 5 | 3092 | |||
BC063143.1 | - | 1 | 1277 | |||
CR749381.1 | - | 1 | 2071 | |||
CR936599.1 | - | 4 | 2943 | |||
CR936695.1 | - | 4 | 2947 |