Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145232.3 | NP_660275.2 | 2 | 497 | ||
GenBank mRNA | AK291173.1 | - | 2 | 477 | ||
BC009037.2 | - | 1 | 453 | 4182164 | ||
BX440723.2 | - | 2 | 497 |